Learn More
Apexbio Technology LLC FERROZINE-500MG
SDP
Catalog No. 5000569378
Encompass_Preferred
Ferrozine (CAS No 69898-45-9) is a chemical compound commonly used in biomedical research especially in studies involving metal ion interactions Originating as an analytical reagent its primary mechanism of action involves chelating iron ions forming a stable complex that allows for quantitative spectrophotometric measurement This property makes it particularly valuable in evaluating iron levels in various biological and environmental samples Its in vitro activity is typically observed in the nanomolar to low micromolar range indicating high sensitivity and specificity for iron detection Ferrozine is frequently employed in cell culture models animal studies and drug screening assays to investigate iron metabolism and related pathways Experimental concentrations typically depend on the specific research design ensuring adaptability across various experimental setups It serves as an essential tool for researchers aiming to understand iron-related biological processes
Catalog No. 50-005-69378 Supplier Apexbio Technology LLC Supplier No. C8217500
Ferrozine (CAS No 69898-45-9) is a chemical compound commonly used in biomedical research especially in studies involving metal ion interactions Originating as an analytical reagent its primary mechanism of action involves chelating iron ions forming a stable complex that allows for quantitative spectrophotometric measurement This property makes it particularly valuable in evaluating iron levels in various biological and environmental samples Its in vitro activity is typically observed in the nanomolar to low micromolar range indicating high sensitivity and specificity for iron detection Ferrozine is frequently employed in cell culture models animal studies and drug screening assays to investigate iron metabolism and related pathways Experimental concentrations typically depend on the specific research design ensuring adaptability across various experimental setups It serves as an essential tool for researchers aiming to understand iron-related biological processes