Learn More
Medchemexpress LLC Ferrozine | 69898-45-9 | 99.9% | 492.46 | 500 MG
SDP
Catalog No. 5000351444
Encompass_Preferred
Ferrozine is a spectrophotometric reagent for iron ions, reacting with divalent Fe to form a stable magenta complex with an absorption peak at 562 nm. Ferrozine-based colorimetric assays can be used to quantify iron in cells.
  • Spectrophotometric reagent for iron ions
  • Reacts with divalent Fe to form stable magenta complex
  • Absorption peak at 562 nm
  • Suitable for quantifying iron in cells
  • Intended for research use only
Catalog No. 50-003-51444 Supplier Medchemexpress LLC Supplier No. HY137805500MG
Ferrozine is a spectrophotometric reagent for iron ions, reacting with divalent Fe to form a stable magenta complex with an absorption peak at 562 nm. Ferrozine-based colorimetric assays can be used to quantify iron in cells.
  • Spectrophotometric reagent for iron ions
  • Reacts with divalent Fe to form stable magenta complex
  • Absorption peak at 562 nm
  • Suitable for quantifying iron in cells
  • Intended for research use only