Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Lange™ FerroZine™ Iron Reagent Solution Pillows

Catalog No. 501994756 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4756 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-4756 Supplier Hach Lange™ Supplier No. 230166
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hach Lange FerroZine Iron Reagent Solution Pillows, Quantity: 25mL, Method: FerroZine

TRUSTED_SUSTAINABILITY

Specifications

Quantity 25 mL
Test Method FerroZine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.