Learn More
Hach Company FerroZine Iron Reagent, 500 mL
Supplier:  Hach Company 230149
Encompass
For determination of trace levels of iron by the Ferrozine method. Hach Method 8147. Range: 0.009 to 1.40 mg/L. Sample: 25 mL. 500 mL bottle.
Catalog No. 50-199-4755