Learn More
Crescent Chemical Co Inc FERROZINE ANALYTICAL GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE21326.02
Encompass_Preferred
Ferrozine analytical grade Specific reagent for serum iron determination with high sensitivity. For automated instrumental analysis (1, 2). For determination of acidity of urine and serum (3). Size - 5G Storage Conditions - +15 C TO +30 C C
Catalog No. 50-247-588