Learn More
Cayman Chemical 3,3′-[3-(2-Pyridinyl)-1,2,4-triazine-5,6-diyl]bis[benzenesulfonic acid] (sodium salt), Purity: ≥98% (mixture of isomers), CAS: 1353005-64-7, 10 g
SDP
Catalog No. 502991760
Encompass_Preferred
A colorimetric reagent that can be used in the detection of iron; ferrozine-iron complexes can be quantified by colorimetric detection at 562 nm
Catalog No. 50-299-1760 Supplier Cayman Chemical Supplier No. 3146510 G
A colorimetric reagent that can be used in the detection of iron; ferrozine-iron complexes can be quantified by colorimetric detection at 562 nm