Learn More
Trajan Scientific FERRUL SHIMADZU17A LINER 10/PK
SDP
Catalog No. 502530221
Encompass_Preferred
Trajan Shimadzu Liner O-Rings, Graphite, Sealing Ring, For Use With: Shimadzu 17 A (SPL-17 Injector), Packaging Quantity: 10 Pack, Temperature Max: 450 deg.C
Catalog No. 50-253-0221 Supplier Trajan Scientific Supplier No. 0726007
Trajan Shimadzu Liner O-Rings, Graphite, Sealing Ring, For Use With: Shimadzu 17 A (SPL-17 Injector), Packaging Quantity: 10 Pack, Temperature Max: 450 deg.C