Learn More
Waters Corp Waters Ferrule, 1/16, Annealed
Supplier: Waters Corp WAT022330
The Waters Ferrule, 1/16, Annealed is engineered for robust connections in various laboratory applications. This ferrule is specifically designed to facilitate the connection between tubing and Viridis columns.
- Features a 1/16 in size.
- Annealed for optimized flexibility and sealing.
- Designed for connecting tubing to Viridis columns.
- Packaged individually for convenience.