Learn More
Glass Expansion Ferrule 1/16 for VeeSpray nebulizer
SDP
Catalog No. 502703085
Encompass_Preferred
Ferrule 1/16 for VeeSpray Nebulizer (PKT 10). Package of ten replacement 1/16” ferrules for VeeSpray Nebulizer. Suitable for use with Ratchet Connector 70-803-1350, and 1/16” OD capillary sample tubing STT-16-75 to make custom length sample line to VeeSpray Nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3085 Supplier Glass Expansion Supplier No. 708030780
Ferrule 1/16 for VeeSpray Nebulizer (PKT 10). Package of ten replacement 1/16” ferrules for VeeSpray Nebulizer. Suitable for use with Ratchet Connector 70-803-1350, and 1/16” OD capillary sample tubing STT-16-75 to make custom length sample line to VeeSpray Nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001