Learn More
Waters Corp Ferrule, ESI GVF 16-16, 2 pc/PAK
Supplier:  Waters Corp 700000343
Encompass_Preferred
This product is an ESI GVF 16-16 ferrule, supplied in a pack of two pieces.
  • GVF ferrule type.
  • Packaged as 2 pieces per pack.
  • Compatible with 3100 Mass Detector, EMD 1000 Mass Spectrometer, Q-Tof Premier Mass Spectrometer, SQ Detector, SYNAPT G2 HDMS System, SYNAPT G2 MS System, Synapt High Definition MS, SYNAPT MS, TQ Detector, XEVO Q-Tof Mass Spec, XEVO.
Catalog No. 50-466-563