Learn More
Microsolv Technology Corporation Ferrule Etfe For 1/16 Ff Nut
Catalog No. 5001451490
Encompass_Preferred
Ferrules, for use with Low Pressure, Tubing Connectors. Use with Flange Free, 1/16th, low pressure tubing. ETFE. MicroSolv Brand. 10/PK.
Catalog No. 50-014-51490 Supplier Microsolv Technology Corporation Supplier No. 45216FETFE
Ferrules, for use with Low Pressure, Tubing Connectors. Use with Flange Free, 1/16th, low pressure tubing. ETFE. MicroSolv Brand. 10/PK.