Learn More
Chemglass Life Sciences Ferrule, Flangeless, 1/16", Blue, Tefzel
SDP
Catalog No. 501531027
Encompass_Preferred
Ferrule, Flangeless, 1/16", Blue, Tefzel. Used with flangeless nut to ensure the tubing is held firmly in place. Ferrule designed to form a zero dead-volume connection. Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.
Catalog No. 50-153-1027 Supplier Chemglass Life Sciences Supplier No. CG1164F01
Ferrule, Flangeless, 1/16", Blue, Tefzel. Used with flangeless nut to ensure the tubing is held firmly in place. Ferrule designed to form a zero dead-volume connection. Ferrule is placed on the end of the tubing with the cone pointing toward the nut, flat end towards the PTFE fitting.