Learn More
IDEX Health & Science Upchurch FERRULE FLANGLSS 2MM OD 10/PK
Catalog No. NC1919061
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, Tefzel, Green, For Use With: 1.8 to 2 mm O.D. Tubing, Length: 0.19 in., Width: 0.19 in., Flangeless Ferrule, ETFE.
Catalog No. NC1919061 Supplier IDEX Health & Science Upchurch Supplier No. P342X
Idex Flangeless Fittings: Ferrules, Tefzel, Green, For Use With: 1.8 to 2 mm O.D. Tubing, Length: 0.19 in., Width: 0.19 in., Flangeless Ferrule, ETFE.