Learn More
IDEX Health & Science Upchurch FERRULE FLNGLSS YWL 1/8IN 10PK
Catalog No. NC1919041
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/8 in. O.D. Tubing, Length: Not Applicable, Width: Not Applicable, Flangeless Ferrule, ETFE.
Catalog No. NC1919041 Supplier IDEX Health & Science Upchurch Supplier No. P300X
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/8 in. O.D. Tubing, Length: Not Applicable, Width: Not Applicable, Flangeless Ferrule, ETFE.