Learn More
Trajan Scientific FERRULE GFS008 SHIMADZU 10PK
SDP
Catalog No. 502530219
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, Reducing Ferrule, Diameter Inside: 0.031 in., 0.08 cm, For Use With: GC, GC14 A, GC17 A, GC2010 and GC2014 Column I.D. 0.45 to 0.53 mm Detector/Injectors, Temperature Maximum: 842 deg.F, Temperature Max: 450 deg.C
Catalog No. 50-253-0219 Supplier Trajan Scientific Supplier No. 0726082
Trajan SGE 100% Graphite Ferrules, Reducing Ferrule, Diameter Inside: 0.031 in., 0.08 cm, For Use With: GC, GC14 A, GC17 A, GC2010 and GC2014 Column I.D. 0.45 to 0.53 mm Detector/Injectors, Temperature Maximum: 842 deg.F, Temperature Max: 450 deg.C