Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 072621
Encompass_Preferred
Graphite ferrule with 1/4 hole for 1/4 OD packed columns PK10
Catalog No. 50-004-94354