Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Catalog No. 5000494507
Encompass_Preferred
Graphite ferrule with 0 8 mm hole for Carlo Erba detector/injectors
Catalog No. 50-004-94507 Supplier Trajan Scientific Supplier No. 072614
Graphite ferrule with 0 8 mm hole for Carlo Erba detector/injectors