Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 072622
Encompass_Preferred
Graphite ferrule with 1/8 hole for 1/8 OD packed columns PK10
Catalog No. 50-004-94916