Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 072624
Encompass_Preferred
Graphite ferrules with 0 5 mm hole for use with 1/8 nuts on PerkinElmer injectors and detectors PK10
Catalog No. 50-004-94424