Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Catalog No. 5000494914
Encompass_Preferred
Graphite ferrule with 0 5 mm hole for Carlo Erba detector/injectors
Catalog No. 50-004-94914 Supplier Trajan Scientific Supplier No. 072619
Graphite ferrule with 0 5 mm hole for Carlo Erba detector/injectors