Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 0726001
Encompass_Preferred
Graphite sealing ring with 5 mm hole for Shimadzu 14 15A and 16 (Spl-14 injector) liners and 5 mm OD packed columns PK10
Catalog No. 50-004-94891