Learn More
Trajan Scientific Ferrule Graphite Medical Lab 10/PK
SDP
Supplier:  Trajan Scientific 0726217
Encompass_Preferred
Graphite sealing ring for Varian 1078 and 1079 injectors PK10
Catalog No. 50-004-93911