Learn More
Trajan Scientific Ferrule Graphite Sealing Ring 10/PK
SDP
Catalog No. 5000494347
Encompass_Preferred
Graphite sealing ring for 3 mm OD PerkinElmer liner PK10
Catalog No. 50-004-94347 Supplier Trajan Scientific Supplier No. 0726522
Graphite sealing ring for 3 mm OD PerkinElmer liner PK10