Learn More
Trajan Scientific Ferrule Graphite/Vespel GV long 10/PK
SDP
Catalog No. 5000494907
Encompass_Preferred
1/16 GC ferrule 1/16 ID (long) 85% Vespel / 15% graphite PK10
Catalog No. 50-004-94907 Supplier Trajan Scientific Supplier No. 215100
1/16 GC ferrule 1/16 ID (long) 85% Vespel / 15% graphite PK10