Learn More
Trajan Scientific Ferrule Graphite/Vespel GV Medical 10/PK
SDP
Catalog No. 5000493696
Encompass_Preferred
15% Graphite/85% Vespel mini union ferrule with 0 4 mm hole PK10
Catalog No. 50-004-93696 Supplier Trajan Scientific Supplier No. 072696
15% Graphite/85% Vespel mini union ferrule with 0 4 mm hole PK10