Learn More
Trajan Scientific Ferrule Graphite/Vespel GV no hole 10/PK
SDP
Catalog No. 5000493783
Encompass_Preferred
1/16 GC ferrule (no-hole) Agilent Style 85% Vespel / 15% graphite PK10
Catalog No. 50-004-93783 Supplier Trajan Scientific Supplier No. 21511X
1/16 GC ferrule (no-hole) Agilent Style 85% Vespel / 15% graphite PK10