Learn More
IDEX Health & Science Upchurch FERRULE LITETOUCH 1/16IN 10PK
Catalog No. NC1919412
Encompass_Preferred
Idex LiteTouch Ferrule, Natural PEEK/SS, 1/16 in. OD Tubing, 10-32 Coned, Description: Fits over 1/16 in. O.D. tubing, For Use With: LiteTouch Ferrule System
Catalog No. NC1919412 Supplier IDEX Health & Science Upchurch Supplier No. LT100X
Idex LiteTouch Ferrule, Natural PEEK/SS, 1/16 in. OD Tubing, 10-32 Coned, Description: Fits over 1/16 in. O.D. tubing, For Use With: LiteTouch Ferrule System