Learn More
Waters Corp Waters Corporation Ferrule, PEEK, 1/16 in., High Pressure
Catalog No. NC9600228 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 1/16 in. ferrule is manufactured from PEEK, designed for high-pressure applications. It is commonly found on many tubing assemblies and primarily utilized in UPLC systems.
  • Manufactured from PEEK material.
  • Suitable for 1/16 in. tubing assemblies.
  • Designed for use in UPLC and HPLC systems.
  • Functions as a component in ACQUITY UPLC Column Managers, ACQUITY UPLC Column Heaters, and 2414 Refractive Index Detectors.
  • Supplied as a package of 10 ferrules.
  • Recommended replacement as required for optimal system performance.
Catalog No. NC9600228 Supplier Waters Corp Supplier No. 700003114
This 1/16 in. ferrule is manufactured from PEEK, designed for high-pressure applications. It is commonly found on many tubing assemblies and primarily utilized in UPLC systems.
  • Manufactured from PEEK material.
  • Suitable for 1/16 in. tubing assemblies.
  • Designed for use in UPLC and HPLC systems.
  • Functions as a component in ACQUITY UPLC Column Managers, ACQUITY UPLC Column Heaters, and 2414 Refractive Index Detectors.
  • Supplied as a package of 10 ferrules.
  • Recommended replacement as required for optimal system performance.