Learn More
Microsolv Technology Corporation Ferrule Puller Tool Blueferru
Catalog No. 5001451501
Encompass_Preferred
Ferrule Puller. Removal Tool for Stainless Steel Ferrules. MicroSolv Brand. 1/EA.
Catalog No. 50-014-51501 Supplier Microsolv Technology Corporation Supplier No. 46026FP
Ferrule Puller. Removal Tool for Stainless Steel Ferrules. MicroSolv Brand. 1/EA.