Learn More
IDEX Health & Science Upchurch FERRULE REPLACE 10-32 10/PK
Catalog No. NC1919742
Encompass_Preferred
Idex Fingertight Ferrules, For Use With: 0.0625 in. O.D. Tubing, PEEK, Natural
Catalog No. NC1919742 Supplier IDEX Health & Science Upchurch Supplier No. F142X
Idex Fingertight Ferrules, For Use With: 0.0625 in. O.D. Tubing, PEEK, Natural