Learn More
Waters Corp Ferrule Set, 0.062 in., 2-Piece, Gold Plated
Catalog No. 502392189 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ferrule set is designed for reliable and secure connections in various analytical applications. It features a durable construction with a gold-plated finish for enhanced performance and longevity. The set includes two pieces, ensuring a complete connection.
  • Diameter: 0.062 in.
  • Configuration: 2-piece
  • Material: Gold plated
Catalog No. 50-239-2189 Supplier Waters Corp Supplier No. 700009325
This ferrule set is designed for reliable and secure connections in various analytical applications. It features a durable construction with a gold-plated finish for enhanced performance and longevity. The set includes two pieces, ensuring a complete connection.
  • Diameter: 0.062 in.
  • Configuration: 2-piece
  • Material: Gold plated