Learn More
Waters Corp Waters Ferrule Set, 1/16 in ID, Two-piece, Stainless Steel
Catalog No. NC9440954 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This spare part is a ferrule set designed for high-performance liquid chromatography systems. It is a quality part suitable for various analytical applications.
  • Type: Fitting
  • Units per package: 10 pk
  • System compatibility: UPC2, HPLC, UPLC
  • Material: Stainless Steel
  • Internal diameter: 1/16 in (1.5875 mm)
Catalog No. NC9440954 Supplier Waters Corp Supplier No. 700002635
This spare part is a ferrule set designed for high-performance liquid chromatography systems. It is a quality part suitable for various analytical applications.
  • Type: Fitting
  • Units per package: 10 pk
  • System compatibility: UPC2, HPLC, UPLC
  • Material: Stainless Steel
  • Internal diameter: 1/16 in (1.5875 mm)