Learn More
IDEX Health & Science Upchurch FERRULE SUP FLNGLSS 1/16 10/PK
Catalog No. NC1919749
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, PEEK, Length: 0.16 in., Width: 0.18 in., Natural, Flangeless Ferrule, For Use With: 1/16 in. O.D. Tubing
Catalog No. NC1919749 Supplier IDEX Health & Science Upchurch Supplier No. P250X
Idex Flangeless Fittings: Ferrules, PEEK, Length: 0.16 in., Width: 0.18 in., Natural, Flangeless Ferrule, For Use With: 1/16 in. O.D. Tubing