Learn More
IDEX Health & Science Upchurch FERRULE SUPR FLNGLS 10/PK
Catalog No. NC1919713
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/16 in. O.D. Tubing, Length: Not Applicable, Width: Not Applicable, Flangeless Ferrule, ETFE
Catalog No. NC1919713 Supplier IDEX Health & Science Upchurch Supplier No. P259X
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/16 in. O.D. Tubing, Length: Not Applicable, Width: Not Applicable, Flangeless Ferrule, ETFE