Learn More
IDEX Health & Science Upchurch FERRULE SUPR FLNGLS W/LCK RNG
Catalog No. NC1919712
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/16 in. O.D. Tubing, Length: 0.16 in., Width: 0.18 in., Flangeless Ferrule, ETFE
Catalog No. NC1919712 Supplier IDEX Health & Science Upchurch Supplier No. P259
Idex Flangeless Fittings: Ferrules, Tefzel, Yellow, For Use With: 1/16 in. O.D. Tubing, Length: 0.16 in., Width: 0.18 in., Flangeless Ferrule, ETFE