Learn More
IDEX Health & Science Upchurch FERRULE SUPR FLNGLSS 1/8 10/PK
Catalog No. NC1919062
Encompass_Preferred
Idex Flangeless Fittings: Ferrules, Tefzel, Natural, For Use With: 1/8 in. O.D. Tubing, Length: 0.16 in., Width: 0.2 in., Flangeless Ferrule, ETFE.
Catalog No. NC1919062 Supplier IDEX Health & Science Upchurch Supplier No. P350X
Idex Flangeless Fittings: Ferrules, Tefzel, Natural, For Use With: 1/8 in. O.D. Tubing, Length: 0.16 in., Width: 0.2 in., Flangeless Ferrule, ETFE.