Learn More
IDEX Health & Science Upchurch FERRULE UPTIGHT 10/PK
Catalog No. NC1919186
Encompass_Preferred
Idex Stainless Steel Ferrules, For Use With: 0.062 in. O.D. Tubing, Certifications/Compliance: ROHS, Length: 0.15 in., 0.38 cm, Max. Pressure: 20000 psi, Port Style: 10 to 32 coned, Quantity: 10/Pk.
Catalog No. NC1919186 Supplier IDEX Health & Science Upchurch Supplier No. U401X
Idex Stainless Steel Ferrules, For Use With: 0.062 in. O.D. Tubing, Certifications/Compliance: ROHS, Length: 0.15 in., 0.38 cm, Max. Pressure: 20000 psi, Port Style: 10 to 32 coned, Quantity: 10/Pk.