Learn More
Trajan Scientific FERRULE VITON SEALNG RNG 10/PK
SDP
Catalog No. 502530212
Encompass_Preferred
Trajan GC Inlet Liner Accessories, Viton O-ring for Agilent GC's, For Use With: Agilent, Thermo trace 1300 Bruker (Varian) Except SG092235, Viton O-Ring, Quantity: 10/Pk., Temperature Max: 300 deg.C
Catalog No. 50-253-0212 Supplier Trajan Scientific Supplier No. 0726532
Trajan GC Inlet Liner Accessories, Viton O-ring for Agilent GC's, For Use With: Agilent, Thermo trace 1300 Bruker (Varian) Except SG092235, Viton O-Ring, Quantity: 10/Pk., Temperature Max: 300 deg.C