Learn More
Microsolv Technology Corporation Ferrules, for use with Two Piece, High Pressure, Tubing Connectors for 1/16" PEEK Tubing. Universal Double Cone. 10/PK.
Catalog No. 502271503
Encompass_Preferred
This Double Cone Ferrules will swage on to the PEEK tubing in two places for a great grip on the tubing. Also, when the MicroSolv Brand 2 Piece PEEK Tubing Connector is used, the shape of this ferrule will not twist the tubing when you turn the fittings.Use with any PEEK, 2 Piece, 1/16th" Tubing Connectors (Male Nuts).
This item is not returnable. View return policy
Catalog No. 50-227-1503 Supplier Microsolv Technology Corporation Supplier No. 4732035
This Double Cone Ferrules will swage on to the PEEK tubing in two places for a great grip on the tubing. Also, when the MicroSolv Brand 2 Piece PEEK Tubing Connector is used, the shape of this ferrule will not twist the tubing when you turn the fittings.Use with any PEEK, 2 Piece, 1/16th" Tubing Connectors (Male Nuts).