Learn More
Microsolv Technology Corporation Ferrules, for use with Low Pressure, Tubing Connectors. Use with Flange Free, 1/8th", low pressure tubing. ETFE. 10/PK.
Catalog No. 502271456
Encompass_Preferred
Use with Endure Brand "Male Nuts" for low pressure tubing connections in HPLC or other fluidic connections. ETFE (Ethylene tetrafluoroethylene) is a fluorine-based plastic. It was designed to have high corrosion resistance and strength over a wide temperature range. ETFE has a relatively high melting temperature, excellent chemical resistance properties.
This item is not returnable. View return policy
Catalog No. 50-227-1456 Supplier Microsolv Technology Corporation Supplier No. 4730302
Use with Endure Brand "Male Nuts" for low pressure tubing connections in HPLC or other fluidic connections. ETFE (Ethylene tetrafluoroethylene) is a fluorine-based plastic. It was designed to have high corrosion resistance and strength over a wide temperature range. ETFE has a relatively high melting temperature, excellent chemical resistance properties.