Learn More
Trajan Scientific FERRULES 10/PK
SDP
Catalog No. 502530216
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, Reducing Ferrule, For Use With: GC, 0.0625 in. Nut, Perkin Elmer and Varian Instruments, Diameter Inside: 0.031 in., 0.08 cm, Length: 0.37 cm, Quantity: 10/Pk., Temperature Max: 450 deg.C, Width: 0.012 in., 0.032 cm
Catalog No. 50-253-0216 Supplier Trajan Scientific Supplier No. 072626
Trajan SGE 100% Graphite Ferrules, Reducing Ferrule, For Use With: GC, 0.0625 in. Nut, Perkin Elmer and Varian Instruments, Diameter Inside: 0.031 in., 0.08 cm, Length: 0.37 cm, Quantity: 10/Pk., Temperature Max: 450 deg.C, Width: 0.012 in., 0.032 cm