Learn More
Trajan Scientific FERRULES 10/PK
SDP
Supplier:  Trajan Scientific 072610
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, Reducing Ferrule, Diameter Inside: 0.019 in., 0.05 cm, For Use With: GC, For Gas Chromatography, Length: 0.032 cm, Temperature Max: 450 deg.C
Catalog No. 50-253-0218