Learn More
Gerstel FERRULES 3D GLASS 10/PK
SDP
Catalog No. NC9531653
Encompass
GP-3D ferrules for CIS 4/6 glass inlet liners (also used with CIS 4/6/TDU liners), Package of 10 GP-3D ferrules for CIS 4/6 glass inlet liners (also used with CIS 4/6/TDU liners), Package of 10
Catalog No. NC9531653 Supplier Gerstel Supplier No. 00754101000
GP-3D ferrules for CIS 4/6 glass inlet liners (also used with CIS 4/6/TDU liners), Package of 10 GP-3D ferrules for CIS 4/6 glass inlet liners (also used with CIS 4/6/TDU liners), Package of 10