Learn More
MilliporeSigma™ Supelco™ Ferrules/Nuts for Visiprep™ Large Volume Sampler
Catalog No. 111005128 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
4/Pk.
Catalog No. Quantity
11-100-5128 4/Pk.
1 options
Catalog No. 11-100-5128 Supplier MilliporeSigma™ Supelco™ Supplier No. 57277
Add to Cart
Add to Cart
Ferrules/Nuts for Visiprep Large Volume Sampler are accessories for use with Visiprep large volume samplers which enable the transfer of low viscosity samples directly from any sample container to conventional solid phase extraction (SPE) tubes on the Visiprep SPE vacuum manifolds.
TRUSTED_SUSTAINABILITY
Specifications
Description Color-Coded
For Use With (Application) Food and Beverages
For Use With (Equipment) Visiprep Large Volume Sampler
Product Line Visiprep
Type SPE Accessory
Quantity 4/Pk.
Visiprep is a trademark of Sigma-Aldrich Co. LLC