Learn More
Microsolv Technology Corporation Ferrules, Stainless Steel, Two Step, Single Cone, 1/16". 10/PK.
Catalog No. 502271585
Encompass_Preferred
These 316L, Stainless Steel Two Step Ferrules are made to be used with 316L, Stainless Steel Tubing Connector, Male Nuts for connecting 1/16th inch SS tubing to injectors, in high pressures zones of HPLC systems. The unique shape of this Ferrule is designed to be used with Rheodyne™ Injectors that require Ferrules like this.
This item is not returnable. View return policy
Catalog No. 50-227-1585 Supplier Microsolv Technology Corporation Supplier No. 49590F1162
These 316L, Stainless Steel Two Step Ferrules are made to be used with 316L, Stainless Steel Tubing Connector, Male Nuts for connecting 1/16th inch SS tubing to injectors, in high pressures zones of HPLC systems. The unique shape of this Ferrule is designed to be used with Rheodyne™ Injectors that require Ferrules like this.