Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fertilysin, MP Biomedicals™

Catalog No. ICN15805091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15805091 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15805091 Supplier MP Biomedicals, Inc Supplier No. 0215805091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

White to off-white powder used for biochemical research.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1477-57-2
Molecular Formula C12H20Cl4N2O2
Molecular Weight (g/mol) 366.104
InChI Key FAOMZVDZARKPFJ-UHFFFAOYSA-N
Synonym fertilysin, bis-diamine, n,n'-octane-1,8-diyl bis 2,2-dichloroacetamide, fertilysine, n,n'-octamethylenebis dichloroacetamide, bisdiamine, fertilysine n, n,n'-bis dichloroacetyl-1,8-octamethylenediamine, n,n'-octamethylenebis 2,2-dichloroacetamide, n,n'-bis dichloracetyl-1,8-octanediamine
PubChem CID 15134
IUPAC Name 2,2-dichloro-N-[8-[(2,2-dichloroacetyl)amino]octyl]acetamide
SMILES C(CCCCNC(=O)C(Cl)Cl)CCCNC(=O)C(Cl)Cl

Specifications

Quantity 1 g
Formula Weight 366.1
Percent Purity 95%
Physical Form Solid
Chemical Name or Material N, N'-Octamethylen-bis(2, 2-Dichloroacetamide)
Recommended Storage Store at 2° to 8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.