Learn More
Cosmo Bio Usa Inc FERTIUP PM 0.5ML-CARD Medium Set of 2
SDP
Catalog No. NC1334440
Encompass
To stabilise and improve spermatozoa cryopreservation results and in vitro fertilization rates for laboratory mice Sperm cryopreservation is a practical and cost-effective method of archiving mouse strains in a laboratory. FERTIUP® and CARD MEDIUM are high-performance media for mouse sperm cryopreservation, sperm preincubation and in vitro fertilization (IVF). When used in combination, FERTIUP® and CARD MEDIUM provide a high and stable rate of fertilization from frozen-thawed mouse sperm, which will allow you to reproduce mice as and when you need them.
Catalog No. NC1334440 Supplier Cosmo Bio Usa Inc Supplier No. KYD005EX
To stabilise and improve spermatozoa cryopreservation results and in vitro fertilization rates for laboratory mice Sperm cryopreservation is a practical and cost-effective method of archiving mouse strains in a laboratory. FERTIUP® and CARD MEDIUM are high-performance media for mouse sperm cryopreservation, sperm preincubation and in vitro fertilization (IVF). When used in combination, FERTIUP® and CARD MEDIUM provide a high and stable rate of fertilization from frozen-thawed mouse sperm, which will allow you to reproduce mice as and when you need them.