Learn More
Cosmo Bio Usa Inc FERTIUP MOUSE SPERM PROTECT
SDP
Catalog No. NC1234267
Encompass
To stabilise and improve spermatozoa cryopreservation results and in vitro fertilization rates for laboratory mice
Catalog No. NC1234267 Supplier Cosmo Bio Usa Inc Supplier No. KYD002EX
To stabilise and improve spermatozoa cryopreservation results and in vitro fertilization rates for laboratory mice