Learn More
Cayman Chemical Ferulenol, Purity: ≥95%, CAS: 6805-34-1, 10 mg
SDP
Catalog No. 502984838
Encompass_Preferred
A prenylated 4-hydroxycoumarin with potent activity against mycobacteria; stimulates tubulin polymerization and is cytotoxic to various human tumor cell lines; disrupts oxidative phosphorylation, inhibiting succinate ubiquinone reductase
Catalog No. 50-298-4838 Supplier Cayman Chemical Supplier No. 1956810 MG
A prenylated 4-hydroxycoumarin with potent activity against mycobacteria; stimulates tubulin polymerization and is cytotoxic to various human tumor cell lines; disrupts oxidative phosphorylation, inhibiting succinate ubiquinone reductase