Learn More
Trajan Scientific FERULE GRAPH 3/8IN GFF3/8 10PK
SDP
Catalog No. 502530217
Encompass_Preferred
Trajan SGE 100% Graphite Ferrules, Straight Ferrule, For Use With: For Gas Chromatography, Temperature Maximum: 842 deg.F, Temperature Max: 450 deg.C
Catalog No. 50-253-0217 Supplier Trajan Scientific Supplier No. 072615
Trajan SGE 100% Graphite Ferrules, Straight Ferrule, For Use With: For Gas Chromatography, Temperature Maximum: 842 deg.F, Temperature Max: 450 deg.C